accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 3232-39-1

IUPAC Name: 1-(acetylsulfanyl)ethan-1-one
Synonyms: 1-(acetylsulfanyl)ethanone
Molecular Weight (g/mol): 118.15
Molecular Formula: C4H6O2S
InChi Key: DPCQJLQPDJPRCM-UHFFFAOYSA-N
SMILES: CC(=O)SC(C)=O
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results