accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOOHOOCH3

CAS RN 32328-03-3

IUPAC Name: 1,5-diethyl 3-hydroxypentanedioate
Synonyms: 1,5-diethyl 3-hydroxypentanedioate
Molecular Weight (g/mol): 204.22
Molecular Formula: C9H16O5
InChi Key: OLLQYIBTJXUEEX-UHFFFAOYSA-N
SMILES: CCOC(=O)CC(O)CC(=O)OCC
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results