accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 32343-73-0

IUPAC Name: N-(5-aminopentyl)acetamide
Synonyms: N-acetylcadaverine
Molecular Weight (g/mol): 144.22
Molecular Formula: C7H16N2O
InChi Key: RMOIHHAKNOFHOE-UHFFFAOYSA-N
SMILES: CC(=O)NCCCCCN

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results