accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOCH3CH2

CAS RN 32360-05-7

IUPAC Name: octadecyl 2-methylprop-2-enoate
Synonyms: octadecyl 2-methylprop-2-enoate
Molecular Weight (g/mol): 338.58
Molecular Formula: C22H42O2
InChi Key: HMZGPNHSPWNGEP-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCCCCCCCCOC(=O)C(C)=C
Boiling Point 
  • (2)
  • (2)
Molecular Weight (g/mol) 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)