accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 32364-72-0

IUPAC Name: [2,2'-bithiophene]-5,5'-dicarbaldehyde
Synonyms: [2,2'-bithiophene]-5,5'-dicarbaldehyde
Molecular Weight (g/mol): 222.28
Molecular Formula: C10H6O2S2
InChi Key: RXAXZMANGDHIJX-UHFFFAOYSA-N
SMILES: O=CC1=CC=C(S1)C1=CC=C(S1)C=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results