accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 324-41-4

IUPAC Name: 2-bromo-6-fluoronaphthalene
Synonyms: 2-bromo-6-fluoronaphthalene
Molecular Weight (g/mol): 225.06
Molecular Formula: C10H6BrF
InChi Key: CAJAZWLCCNCVEY-UHFFFAOYSA-N
SMILES: FC1=CC=C2C=C(Br)C=CC2=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results