accessibility menu, dialog, popup

UserName

Viewing profile as user:
F

CAS RN 324-74-3

IUPAC Name: 4-fluoro-1,1'-biphenyl
Synonyms: 4-fluorobiphenyl
Molecular Weight (g/mol): 172.2
Molecular Formula: C12H9F
InChi Key: RUYZJEIKQYLEGZ-UHFFFAOYSA-N
SMILES: FC1=CC=C(C=C1)C1=CC=CC=C1
Boiling Point 
  • (4)
  • (1)
Molecular Weight (g/mol) 
  • (1)
  • (4)
Percent Purity 
  • (2)
  • (2)
  • (1)
Quantity 
  • (2)
  • (1)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (4)
  • (1)
  • (1)
  • (4)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)