accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 324028-95-7

IUPAC Name: 6-chloro-4-methylpyridine-2-carboxylic acid
Synonyms: 6-chloro-4-methylpyridine-2-carboxylic acid
Molecular Weight (g/mol): 171.58
Molecular Formula: C7H6ClNO2
InChi Key: IDIRHQNTRMGBPJ-UHFFFAOYSA-N
SMILES: CC1=CC(=NC(Cl)=C1)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results