accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 3243-40-1

IUPAC Name: 3-(4-propoxyphenyl)propanoic acid
Synonyms: 3-(4-propoxyphenyl)propanoic acid
Molecular Weight (g/mol): 208.26
Molecular Formula: C12H16O3
InChi Key: SOFMLHAGBATXNZ-UHFFFAOYSA-N
SMILES: CCCOC1=CC=C(CCC(O)=O)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results