accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 32443-99-5

IUPAC Name: (2S)-2-amino-3-sulfanylpropanoic acid hydrochloride
Synonyms: D-cysteine hydrochloride
Molecular Weight (g/mol): 157.61
Molecular Formula: C3H8ClNO2S
InChi Key: IFQSXNOEEPCSLW-HSHFZTNMSA-N
SMILES: Cl.N[C@H](CS)C(O)=O
Grade 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)