accessibility menu, dialog, popup

UserName

Viewing profile as user:
OOClOHClOHH2OH2OH2OBa

CAS RN 32458-20-1

IUPAC Name: 2,5-dichloro-3,6-dihydroxycyclohexa-2,5-diene-1,4-dione barium trihydrate
Synonyms: chloranilic acid barium trihydrate
Molecular Weight (g/mol): 400.35
Molecular Formula: C6H8BaCl2O7
InChi Key: JFBLKYVZQCFPJE-UHFFFAOYSA-N
SMILES: O.O.O.[Ba].OC1=C(Cl)C(=O)C(O)=C(Cl)C1=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (1)
  • (1)