accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3HOCH3CH3

CAS RN 3247-00-5

IUPAC Name: tris(4-methylphenyl)methanol
Synonyms: tris(4-methylphenyl)methanol
Molecular Weight (g/mol): 302.42
Molecular Formula: C22H22O
InChi Key: DNWQXZDDISHGRM-UHFFFAOYSA-N
SMILES: CC1=CC=C(C=C1)C(O)(C1=CC=C(C)C=C1)C1=CC=C(C)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results