accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NH3CN

CAS RN 3248-05-3

IUPAC Name: 4,7-dimethyl-1,10-phenanthroline
Synonyms: 4,7-dimethyl-o-phenanthroline
Molecular Weight (g/mol): 208.26
Molecular Formula: C14H12N2
InChi Key: JIVLDFFWTQYGSR-UHFFFAOYSA-N
SMILES: CC1=C2C=CC3=C(C)C=CN=C3C2=NC=C1
Percent Purity 
  • (1)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (3)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)