CAS RN 3251-23-8
VeriSpec™ Copper Standard for ICP 1000 ppm in 2% HNO3, Ricca Chemical
CAS: 3251-23-8 Molecular Formula: CuN2O6 Molecular Weight (g/mol): 187.554 InChI Key: XTVVROIMIGLXTD-UHFFFAOYSA-N PubChem CID: 18616 ChEBI: CHEBI:78036 IUPAC Name: copper;dinitrate SMILES: [N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[Cu+2]
VeriSpec™ Copper Standard for ICP/MS 100 ppm in 2% HNO3, Ricca Chemical
CAS: 3251-23-8 Molecular Formula: CuN2O6 Molecular Weight (g/mol): 187.554 InChI Key: XTVVROIMIGLXTD-UHFFFAOYSA-N PubChem CID: 18616 ChEBI: CHEBI:78036 IUPAC Name: copper;dinitrate SMILES: [N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[Cu+2]
VeriSpec™ Copper (Cu) Standard for AAS 1000 ppm in 2% HNO3, Ricca Chemical
CAS: 3251-23-8 Molecular Formula: CuN2O6 Molecular Weight (g/mol): 187.554 InChI Key: XTVVROIMIGLXTD-UHFFFAOYSA-N PubChem CID: 18616 ChEBI: CHEBI:78036 IUPAC Name: copper;dinitrate SMILES: [N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[Cu+2]
Copper Standard, 1.0 ppm Cu in 12.5% (v/v) Ethanol, Ricca Chemical
CAS: 3251-23-8 Molecular Formula: CuN2O6 Molecular Weight (g/mol): 187.554 InChI Key: XTVVROIMIGLXTD-UHFFFAOYSA-N PubChem CID: 18616 ChEBI: CHEBI:78036 IUPAC Name: copper;dinitrate SMILES: [N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[Cu+2]
VeriSpec™ Copper Standard for ICP/MS 10 ppm in 2% HNO3, Ricca Chemical
CAS: 3251-23-8 Molecular Formula: CuN2O6 Molecular Weight (g/mol): 187.554 InChI Key: XTVVROIMIGLXTD-UHFFFAOYSA-N PubChem CID: 18616 ChEBI: CHEBI:78036 IUPAC Name: copper;dinitrate SMILES: [N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[Cu+2]
VeriSpec™ Copper Standard for ICP 10 g/L in 5% HNO3, Ricca Chemical
CAS: 3251-23-8 Molecular Formula: CuN2O6 Molecular Weight (g/mol): 187.554 InChI Key: XTVVROIMIGLXTD-UHFFFAOYSA-N PubChem CID: 18616 ChEBI: CHEBI:78036 IUPAC Name: copper;dinitrate SMILES: [N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[Cu+2]
Copper Standard, 20 ppm Cu, Ricca Chemical
CAS: 3251-23-8 Molecular Formula: CuN2O6 Molecular Weight (g/mol): 187.554 InChI Key: XTVVROIMIGLXTD-UHFFFAOYSA-N PubChem CID: 18616 ChEBI: CHEBI:78036 IUPAC Name: copper;dinitrate SMILES: [N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[Cu+2]
Copper Standard, 0.3 ppm Cu in 12.5% (v/v) Ethanol, Ricca Chemical
CAS: 3251-23-8 Molecular Formula: CuN2O6 Molecular Weight (g/mol): 187.554 InChI Key: XTVVROIMIGLXTD-UHFFFAOYSA-N PubChem CID: 18616 ChEBI: CHEBI:78036 IUPAC Name: copper;dinitrate SMILES: [N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[Cu+2]
Ricca Chemical Company Copper Standard, 1mL = 1mg Cu, 1000ppm Cu, Ricca Chemical
CAS: 3251-23-8 Molecular Formula: CuN2O6 Molecular Weight (g/mol): 187.554 InChI Key: XTVVROIMIGLXTD-UHFFFAOYSA-N PubChem CID: 18616 ChEBI: CHEBI:78036 IUPAC Name: copper;dinitrate SMILES: [N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[Cu+2]