accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OOOHO

CAS RN 32529-79-6

IUPAC Name: 4-(methoxycarbonyl)cyclohexane-1-carboxylic acid
Synonyms: 4-(methoxycarbonyl)cyclohexane-1-carboxylic acid
Molecular Weight (g/mol): 186.21
Molecular Formula: C9H14O4
InChi Key: ZQJNPHCQABYENK-UHFFFAOYSA-N
SMILES: COC(=O)C1CCC(CC1)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results