accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 3253-39-2

IUPAC Name: 4-(2-{4-[(2-methylprop-2-enoyl)oxy]phenyl}propan-2-yl)phenyl 2-methylprop-2-enoate
Synonyms: bisphenol A dimethacrylate Bis-DMA
Molecular Weight (g/mol): 364.44
Molecular Formula: C23H24O4
InChi Key: QUZSUMLPWDHKCJ-UHFFFAOYSA-N
SMILES: CC(=C)C(=O)OC1=CC=C(C=C1)C(C)(C)C1=CC=C(OC(=O)C(C)=C)C=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results