accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 325810-07-9

IUPAC Name: λ¹-copper(1+) trifluoromethanide tris(triphenylphosphane)
Synonyms: λ¹-copper(1+) trifluoromethanide tris(triphenylphosphine)
Molecular Weight (g/mol): 919.43
Molecular Formula: C55H45CuF3P3
InChi Key: ADEZRALRKPUAPZ-UHFFFAOYSA-N
SMILES: [Cu+].F[C-](F)F.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results