accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 326-06-7

IUPAC Name: 4,4,4-trifluoro-1-phenylbutane-1,3-dione
Synonyms: 4,4,4-trifluoro-1-phenylbutane-1,3-dione
Molecular Weight (g/mol): 216.16
Molecular Formula: C10H7F3O2
InChi Key: VVXLFFIFNVKFBD-UHFFFAOYSA-N
SMILES: FC(F)(F)C(=O)CC(=O)C1=CC=CC=C1
Boiling Point 
  • (3)
  • (1)
Percent Purity 
  • (2)
  • (1)
  • (1)
Quantity 
  • (1)
  • (2)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)
  • (2)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)