accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OOOO

CAS RN 326-61-4

IUPAC Name: (2H-1,3-benzodioxol-5-yl)methyl acetate
Synonyms: piperonyl acetate
Molecular Weight (g/mol): 194.19
Molecular Formula: C10H10O4
InChi Key: PFWYHTORQZAGCA-UHFFFAOYSA-N
SMILES: CC(=O)OCC1=CC=C2OCOC2=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results