accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 326-78-3

IUPAC Name: 3-fluoro-4-(octyloxy)benzoic acid
Synonyms: 3-fluoro-4-(octyloxy)benzoic acid
Molecular Weight (g/mol): 268.33
Molecular Formula: C15H21FO3
InChi Key: SQVFTDZEUGEJTQ-UHFFFAOYSA-N
SMILES: CCCCCCCCOC1=C(F)C=C(C=C1)C(O)=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results