accessibility menu, dialog, popup

UserName

Viewing profile as user:
FFFOOO

CAS RN 326-90-9

IUPAC Name: 4,4,4-trifluoro-1-(furan-2-yl)butane-1,3-dione
Synonyms: 4,4,4-trifluoro-1-(furan-2-yl)butane-1,3-dione
Molecular Weight (g/mol): 206.12
Molecular Formula: C8H5F3O3
InChi Key: OWLPCALGCHDBCN-UHFFFAOYSA-N
SMILES: FC(F)(F)C(=O)CC(=O)C1=CC=CO1
Boiling Point 
  • (2)
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)