CAS RN 326-91-0
Thenoyltrifluoroacetone, MP Biomedicals™
CAS: 326-91-0 Molecular Formula: C8H5F3O2S Molecular Weight (g/mol): 222.18 MDL Number: MFCD00005445 InChI Key: TXBBUSUXYMIVOS-UHFFFAOYSA-N Synonym: 2-thenoyltrifluoroacetone,thenoyltrifluoroacetone,1,3-butanedione, 4,4,4-trifluoro-1-2-thienyl,4,4,4-trifluoro-1-thiophen-2-yl butane-1,3-dione,4,4,4-trifluoro-1-2-thienyl-1,3-butanedione,4,4,4-trifluoro-1-2-thienyl butane-1,3-dione,perfluoroacetyl 2-thenoyl methane,alpha-thenoyltrifluoroacetone,1-thenoyl-3,3,3-trifluoroacetone,.alpha.-thenoyltrifluoroacetone PubChem CID: 5601 IUPAC Name: 4,4,4-trifluoro-1-(thiophen-2-yl)butane-1,3-dione SMILES: FC(F)(F)C(=O)CC(=O)C1=CC=CS1
1-(2-Thenoyl)-3,3,3-trifluoroacetone, 99% (dry wt.) may cont. up to ca 2% water
CAS: 326-91-0 Molecular Formula: C8H5F3O2S Molecular Weight (g/mol): 222.18 MDL Number: MFCD00005445 InChI Key: TXBBUSUXYMIVOS-UHFFFAOYSA-N Synonym: 2-thenoyltrifluoroacetone,thenoyltrifluoroacetone,1,3-butanedione, 4,4,4-trifluoro-1-2-thienyl,4,4,4-trifluoro-1-thiophen-2-yl butane-1,3-dione,4,4,4-trifluoro-1-2-thienyl-1,3-butanedione,4,4,4-trifluoro-1-2-thienyl butane-1,3-dione,perfluoroacetyl 2-thenoyl methane,alpha-thenoyltrifluoroacetone,1-thenoyl-3,3,3-trifluoroacetone,.alpha.-thenoyltrifluoroacetone PubChem CID: 5601 IUPAC Name: 4,4,4-trifluoro-1-thiophen-2-ylbutane-1,3-dione SMILES: FC(F)(F)C(=O)CC(=O)C1=CC=CS1
Thenoyltrifluoroacetone, 99%
CAS: 326-91-0 Molecular Formula: C8H5F3O2S Molecular Weight (g/mol): 222.18 MDL Number: MFCD00005445 InChI Key: TXBBUSUXYMIVOS-UHFFFAOYSA-N Synonym: 2-thenoyltrifluoroacetone,thenoyltrifluoroacetone,1,3-butanedione, 4,4,4-trifluoro-1-2-thienyl,4,4,4-trifluoro-1-thiophen-2-yl butane-1,3-dione,4,4,4-trifluoro-1-2-thienyl-1,3-butanedione,4,4,4-trifluoro-1-2-thienyl butane-1,3-dione,perfluoroacetyl 2-thenoyl methane,alpha-thenoyltrifluoroacetone,1-thenoyl-3,3,3-trifluoroacetone,.alpha.-thenoyltrifluoroacetone PubChem CID: 5601 IUPAC Name: 4,4,4-trifluoro-1-thiophen-2-ylbutane-1,3-dione SMILES: FC(F)(F)C(=O)CC(=O)C1=CC=CS1
4,4,4-Trifluoro-1-(2-thienyl)-1,3-butanedione 98.0+%, TCI America™
CAS: 326-91-0 Molecular Formula: C8H5F3O2S Molecular Weight (g/mol): 222.18 MDL Number: MFCD00005445 InChI Key: TXBBUSUXYMIVOS-UHFFFAOYSA-N Synonym: 2-thenoyltrifluoroacetone,thenoyltrifluoroacetone,1,3-butanedione, 4,4,4-trifluoro-1-2-thienyl,4,4,4-trifluoro-1-thiophen-2-yl butane-1,3-dione,4,4,4-trifluoro-1-2-thienyl-1,3-butanedione,4,4,4-trifluoro-1-2-thienyl butane-1,3-dione,perfluoroacetyl 2-thenoyl methane,alpha-thenoyltrifluoroacetone,1-thenoyl-3,3,3-trifluoroacetone,.alpha.-thenoyltrifluoroacetone PubChem CID: 5601 IUPAC Name: 4,4,4-trifluoro-1-(thiophen-2-yl)butane-1,3-dione SMILES: FC(F)(F)C(=O)CC(=O)C1=CC=CS1
Sigma Aldrich 2-Thenoyltrifluoroacetone
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.