CAS RN 3261-87-8
IUPAC Name:
1,4-oxathiane-2,6-dione
Synonyms:
1,4-oxathiane-2,6-dione
Molecular Weight (g/mol):
132.13
Molecular Formula:
C4H4O3S
InChi Key:
RIIUAPMWDSRBSH-UHFFFAOYSA-N
SMILES:
O=C1CSCC(=O)O1
1
–
1
of
1
results
1
Promotions Available
Thiodiglycolic anhydride, 98%
Thiodiglycolic anhydride, 98%
CAS: 3261-87-8 Molecular Formula: C4H4O3S Molecular Weight (g/mol): 132.133 MDL Number: MFCD00051689 InChI Key: RIIUAPMWDSRBSH-UHFFFAOYSA-N Synonym: thiodiglycolic anhydride,1-oxa-4-thia-cyclohexan-2,6-dione PubChem CID: 137871 IUPAC Name: 1,4-oxathiane-2,6-dione SMILES: C1C(=O)OC(=O)CS1