accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 32679-02-0

IUPAC Name: λ¹-rhodium(1+) (1Z,5Z)-cycloocta-1,5-diene bis(acetonitrile) tetrafluoroboranuide
Synonyms: λ¹-rhodium(1+) 1,5-cyclooctadiene, (Z,Z)- bis(acetonitrile) tetrafluoroborate
Molecular Weight (g/mol): 380
Molecular Formula: C12H18BF4N2Rh
InChi Key: KSMAFOANQHYEJG-SUESZSCISA-N
SMILES: [Rh+].CC#N.CC#N.F[B-](F)(F)F.C1C\C=C/CC\C=C/1
Molecular Weight (g/mol) 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results