accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3IH3CCH3ICH3

CAS RN 3268-21-1

IUPAC Name: 1,4-diiodo-2,3,5,6-tetramethylbenzene
Synonyms: 1,4-diiodo-2,3,5,6-tetramethylbenzene
Molecular Weight (g/mol): 386.02
Molecular Formula: C10H12I2
InChi Key: YSKCEXICRGEWDI-UHFFFAOYSA-N
SMILES: CC1=C(C)C(I)=C(C)C(C)=C1I
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results