accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CSO

CAS RN 3268-49-3

IUPAC Name: 3-(methylsulfanyl)propanal
Synonyms: methional methional (natural)
Molecular Weight (g/mol): 104.17
Molecular Formula: C4H8OS
InChi Key: CLUWOWRTHNNBBU-UHFFFAOYSA-N
SMILES: CSCCC=O
Boiling Point 
  • (2)
  • (1)
  • (1)
Molecular Weight (g/mol) 
  • (3)
  • (1)
Percent Purity 
  • (1)
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)