accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3C(S)NH2OHO

CAS RN 327-57-1

IUPAC Name: (2S)-2-aminohexanoic acid
Synonyms: norleucine|L(+)-norleucine|Nle|2-aminocaproic acid
Molecular Weight (g/mol): 131.18
Molecular Formula: C6H13NO2
InChi Key: LRQKBLKVPFOOQJ-YFKPBYRVSA-N
SMILES: CCCC[C@H](N)C(O)=O
Percent Purity 
  • (2)
  • (2)
  • (2)
Quantity 
  • (3)
  • (1)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (4)
  • (2)
  • (2)
  • (2)
  • (3)
  • (1)
  • (2)
  • (2)