accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 327-74-2

IUPAC Name: 4-amino-3-(trifluoromethyl)benzonitrile
Synonyms: 4-amino-3-(trifluoromethyl)benzonitrile
Molecular Weight (g/mol): 186.14
Molecular Formula: C8H5F3N2
InChi Key: MWLZJOBGDXBMBP-UHFFFAOYSA-N
SMILES: NC1=C(C=C(C=C1)C#N)C(F)(F)F
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results