CAS RN 3270-97-1
IUPAC Name:
6-methoxypyrimidine-2,4-diamine
Synonyms:
6-methoxypyrimidine-2,4-diamine
Molecular Weight (g/mol):
140.15
Molecular Formula:
C5H8N4O
InChi Key:
OSBMJXWHJWWZJP-UHFFFAOYSA-N
SMILES:
COC1=NC(N)=NC(N)=C1
1
–
1
of
1
results
1
2,4-Diamino-6-methoxypyrimidine 98.0+%, TCI America™
CAS: 3270-97-1 Molecular Formula: C5H8N4O Molecular Weight (g/mol): 140.146 MDL Number: MFCD02091088 InChI Key: OSBMJXWHJWWZJP-UHFFFAOYSA-N PubChem CID: 816925 IUPAC Name: 6-methoxypyrimidine-2,4-diamine SMILES: COC1=NC(=NC(=C1)N)N