CAS RN 327056-73-5
IUPAC Name:
3-chloro-5-fluorobenzonitrile
Synonyms:
3-chloro-5-fluorobenzonitrile
Molecular Weight (g/mol):
155.56
Molecular Formula:
C7H3ClFN
InChi Key:
GIKLRWNRBOLRDV-UHFFFAOYSA-N
SMILES:
FC1=CC(Cl)=CC(=C1)C#N
1
–
1
of
1
results
1
Sigma Aldrich 3-Chloro-5-fluorobenzonitrile
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.