accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 32707-89-4

IUPAC Name: [3,5-bis(trifluoromethyl)phenyl]methanol
Synonyms: [3,5-bis(trifluoromethyl)phenyl]methanol
Molecular Weight (g/mol): 244.14
Molecular Formula: C9H6F6O
InChi Key: BJTWPJOGDWRYDD-UHFFFAOYSA-N
SMILES: OCC1=CC(=CC(=C1)C(F)(F)F)C(F)(F)F
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (2)
  • (2)
Boiling Point 
  • (3)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (1)
  • (2)
  • (2)
  • (3)
  • (2)