accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 3272-08-0

IUPAC Name: 4-cyano-2-nitrobenzen-1-olate
Synonyms: 4-cyano-2-nitrobenzenolate
Molecular Weight (g/mol): 163.11
Molecular Formula: C7H3N2O3
InChi Key: INBLGVOPOSGVTA-UHFFFAOYSA-M
SMILES: [O-]C1=CC=C(C=C1[N+]([O-])=O)C#N
Molecular Weight (g/mol) 
  • (2)
  • (1)
Quantity 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results