accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOH3C

CAS RN 32723-67-4

IUPAC Name: 4-methoxy-3-methylbenzaldehyde
Synonyms: 3-methyl-4-methoxybenzaldehyde
Molecular Weight (g/mol): 150.18
Molecular Formula: C9H10O2
InChi Key: MYLBIQHZWFWSMH-UHFFFAOYSA-N
SMILES: COC1=CC=C(C=O)C=C1C
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results