accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 32740-79-7

IUPAC Name: dioxoruthenium
Synonyms: ruthenium(IV) oxide
Molecular Weight (g/mol): 133.07
Molecular Formula: O2Ru
InChi Key: WOCIAKWEIIZHES-UHFFFAOYSA-N
SMILES: O=[Ru]=O
Molecular Weight (g/mol) 
  • (10)
  • (1)
Quantity 
  • (4)
  • (3)
  • (4)
Physical Form 
  • (3)
  • (4)
Percent Purity 
  • (3)
  • (1)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (7)
  • (10)
  • (1)
  • (4)
  • (3)
  • (4)
  • (3)
  • (4)
  • (3)
  • (1)
  • (1)