accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3SiH3COSiCH3H3C

CAS RN 3277-26-7

IUPAC Name: —
Synonyms: —
Molecular Weight (g/mol): 132.31
Molecular Formula: C4H12OSi2
InChi Key: KWEKXPWNFQBJAY-UHFFFAOYSA-N
SMILES: C[Si](C)O[Si](C)C
Molecular Weight (g/mol) 
  • (5)
  • (2)
  • (3)
Quantity 
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (2)
  • (6)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (2)
  • (2)
  • (6)
  • (5)
  • (2)
  • (3)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (6)
  • (5)
  • (2)