accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 32771-64-5

IUPAC Name: 2-methyl-4-[(7H-purin-6-yl)amino]but-2-en-1-ol
Synonyms: zeatin
Molecular Weight (g/mol): 219.25
Molecular Formula: C10H13N5O
InChi Key: UZKQTCBAMSWPJD-UHFFFAOYSA-N
SMILES: CC(CO)=CCNC1=C2NC=NC2=NC=N1
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results