accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH2SOOSOOH2C

CAS RN 3278-22-6

IUPAC Name: (ethenesulfonyl)methanesulfonylethene
Synonyms: (ethenesulfonyl)methanesulfonylethene
Molecular Weight (g/mol): 196.24
Molecular Formula: C5H8O4S2
InChi Key: IJHIIHORMWQZRQ-UHFFFAOYSA-N
SMILES: C=CS(=O)(=O)CS(=O)(=O)C=C
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)