accessibility menu, dialog, popup

UserName

Viewing profile as user:
OONNOO

CAS RN 3278-31-7

IUPAC Name: 1-[4-(2,5-dioxo-2,5-dihydro-1H-pyrrol-1-yl)phenyl]-2,5-dihydro-1H-pyrrole-2,5-dione
Synonyms: 1-[4-(2,5-dioxopyrrol-1-yl)phenyl]pyrrole-2,5-dione
Molecular Weight (g/mol): 268.23
Molecular Formula: C14H8N2O4
InChi Key: AQGZJQNZNONGKY-UHFFFAOYSA-N
SMILES: O=C1C=CC(=O)N1C1=CC=C(C=C1)N1C(=O)C=CC1=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)