accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 32780-06-6

IUPAC Name: 5-(hydroxymethyl)oxolan-2-one
Synonyms: 5-(hydroxymethyl)oxolan-2-one
Molecular Weight (g/mol): 116.12
Molecular Formula: C5H8O3
InChi Key: NSISJFFVIMQBRN-UHFFFAOYNA-N
SMILES: OCC1CCC(=O)O1
Molecular Weight (g/mol) 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)