CAS RN 32795-84-9
IUPAC Name:
7-bromotetraphene
Synonyms:
7-bromotetraphene
Molecular Weight (g/mol):
307.19
Molecular Formula:
C18H11Br
InChi Key:
LGRNWCDRODWMOH-UHFFFAOYSA-N
SMILES:
BrC1=C2C=CC3=CC=CC=C3C2=CC2=CC=CC=C12
1
–
1
of
1
results
1
7-Bromobenz[a]anthracene 98.0+%, TCI America™
CAS: 32795-84-9 Molecular Formula: C18H11Br Molecular Weight (g/mol): 307.19 MDL Number: MFCD00029016 InChI Key: LGRNWCDRODWMOH-UHFFFAOYSA-N PubChem CID: 72849 IUPAC Name: 7-bromobenzo[a]anthracene SMILES: C1=CC=C2C(=C1)C=CC3=C(C4=CC=CC=C4C=C32)Br