accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CCH3NH2OHO

CAS RN 328-39-2

IUPAC Name: 2-amino-4-methylpentanoic acid
Synonyms: leucine|leucin|leuzin|Hleu
Molecular Weight (g/mol): 131.18
Molecular Formula: C6H13NO2
InChi Key: ROHFNLRQFUQHCH-UHFFFAOYNA-N
SMILES: CC(C)CC(N)C(O)=O
Percent Purity 
  • (1)
  • (3)
  • (2)
Quantity 
  • (1)
  • (2)
  • (2)
  • (1)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (5)
  • (1)
  • (3)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)