accessibility menu, dialog, popup

UserName

Viewing profile as user:
HOOOOHO

CAS RN 328-42-7

IUPAC Name: 2-oxobutanedioic acid
Synonyms: oxalacetate OAA ion(2-) oxobutanedioic acid oxalacetic acid oxalacetic acetic
Molecular Weight (g/mol): 132.07
Molecular Formula: C4H4O5
InChi Key: KHPXUQMNIQBQEV-UHFFFAOYSA-N
SMILES: OC(=O)CC(=O)C(O)=O
Molecular Weight (g/mol) 
  • (3)
  • (9)
Quantity 
  • (1)
  • (2)
  • (1)
  • (3)
  • (4)
Percent Purity 
  • (2)
  • (2)
  • (3)
  • (3)
Physical Form 
  • (2)
  • (3)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (1)
  • (1)
  • (2)
  • (8)
  • (3)
  • (9)
  • (1)
  • (2)
  • (1)
  • (3)
  • (4)
  • (2)
  • (2)
  • (3)
  • (3)
  • (2)
  • (3)
  • (2)
  • (1)