accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 328-67-6

IUPAC Name: 3-bromo-5-(trifluoromethyl)benzoic acid
Synonyms: 3-bromo-5-(trifluoromethyl)benzoic acid
Molecular Weight (g/mol): 269.02
Molecular Formula: C8H4BrF3O2
InChi Key: AMZBKZQMAZWIJM-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC(=CC(Br)=C1)C(F)(F)F
Percent Purity 
  • (1)
  • (2)
Physical Form 
  • (1)
  • (2)
Quantity 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)