accessibility menu, dialog, popup

UserName

Viewing profile as user:
FFFH2NOOH

CAS RN 328-68-7

IUPAC Name: 3-amino-5-(trifluoromethyl)benzoic acid
Synonyms: 3-amino-5-(trifluoromethyl)benzoic acid
Molecular Weight (g/mol): 205.14
Molecular Formula: C8H6F3NO2
InChi Key: WBTHOSZMTIPJLR-UHFFFAOYSA-N
SMILES: NC1=CC(=CC(=C1)C(F)(F)F)C(O)=O
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (2)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)