accessibility menu, dialog, popup

UserName

Viewing profile as user:
FFFClCl

CAS RN 328-84-7

IUPAC Name: 1,2-dichloro-4-(trifluoromethyl)benzene
Synonyms: 3,4-dichlorobenzotrifluoride
Molecular Weight (g/mol): 215
Molecular Formula: C7H3Cl2F3
InChi Key: XILPLWOGHPSJBK-UHFFFAOYSA-N
SMILES: FC(F)(F)C1=CC=C(Cl)C(Cl)=C1
Molecular Weight (g/mol) 
  • (4)
  • (1)
Boiling Point 
  • (2)
  • (1)
  • (1)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (4)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)