accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 328-93-8

IUPAC Name: 2,5-bis(trifluoromethyl)aniline
Synonyms: 2,5-bis(trifluoromethyl)aniline
Molecular Weight (g/mol): 229.13
Molecular Formula: C8H5F6N
InChi Key: XWMVIJUAZAEWIE-UHFFFAOYSA-N
SMILES: NC1=CC(=CC=C1C(F)(F)F)C(F)(F)F
Boiling Point 
  • (2)
  • (3)
Molecular Weight (g/mol) 
  • (1)
  • (4)
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (2)
  • (3)
  • (1)
  • (4)
  • (2)
  • (3)
  • (1)
  • (2)
  • (2)
  • (2)