accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 328023-30-9

IUPAC Name: N-(4-ethoxyphenyl)-3-methylbenzamide
Synonyms: N-(4-ethoxyphenyl)-3-methylbenzamide
Molecular Weight (g/mol): 255.32
Molecular Formula: C16H17NO2
InChi Key: MMLNIRUCJDCWDS-UHFFFAOYSA-N
SMILES: CCOC1=CC=C(NC(=O)C2=CC(C)=CC=C2)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results