accessibility menu, dialog, popup

UserName

Viewing profile as user:
NH2NH2

CAS RN 3282-99-3

IUPAC Name: 4-[4-(4-aminophenyl)cyclohexyl]aniline
Synonyms: 4-[4-(4-aminophenyl)cyclohexyl]aniline
Molecular Weight (g/mol): 266.39
Molecular Formula: C18H22N2
InChi Key: DJCVCYUEICJZBD-UHFFFAOYSA-N
SMILES: NC1=CC=C(C=C1)C1CCC(CC1)C1=CC=C(N)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results