accessibility menu, dialog, popup

UserName

Viewing profile as user:
NOBr

CAS RN 328270-70-8

IUPAC Name: 5-(2-bromophenyl)-1,3-oxazole
Synonyms: 5-(2-bromophenyl)-1,3-oxazole
Molecular Weight (g/mol): 224.06
Molecular Formula: C9H6BrNO
InChi Key: JLTHLCLAPCIKJJ-UHFFFAOYSA-N
SMILES: BrC1=CC=CC=C1C1=CN=CO1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results