accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 3283-05-4

IUPAC Name: N1,N1,N4,N4-tetrakis(4-nitrophenyl)benzene-1,4-diamine
Synonyms: N1,N1,N4,N4-tetrakis(4-nitrophenyl)benzene-1,4-diamine
Molecular Weight (g/mol): 592.52
Molecular Formula: C30H20N6O8
InChi Key: XEUNCVYZWDLKKR-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC=C(C=C1)N(C1=CC=C(C=C1)N(C1=CC=C(C=C1)[N+]([O-])=O)C1=CC=C(C=C1)[N+]([O-])=O)C1=CC=C(C=C1)[N+]([O-])=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results